Anti-ODF2 Rabbit Polyclonal Antibody

Supplier: Proteintech
12058-1-AP-150UL
10091-548EA 462.47 USD
10091-548
Anti-ODF2 Rabbit Polyclonal Antibody
Antibodies
ODF2 (outer dense fiber 2) is the major cenexin protein of the cytoskeleton of the sperm tail. In somatic cells, it is a component of the centrosome in which it is located in the appendages of the mother centriole. The gene for the cenexin protein, which was first identified as a protein marker of the mother centriole, was subsequently found to be identical to ODF2 and encodes transcripts cenexin 1 and cenexin 2. ODF2 is ubiquitously expressed in many cell types as various splicing variants. Additionally, ODF2 localizes to centrioles, basal bodies, and primary cilia, which are all structurally and functionally interconnected. ODF2 has 10 isoforms with MW 67-100 kDa.

Western Blot: Mouse Testis Tissue, 1:200-1:2000; IHC: Human breast cancer Tissue, 1:20-1:200; IP: Mouse Testis Tissue, 1:200-1:2000

Type: Primary
Antigen: ODF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR