Anti-OCTN2 Rabbit Polyclonal Antibody

Supplier: Proteintech
16331-1-AP-150UL
10091-544EA 462.47 USD
10091-544
Anti-OCTN2 Rabbit Polyclonal Antibody
Antibodies
OCTN2 (Organic Cation/Carnitine Transporter 2), encoded by SLC22A5, is a ubiquitously expressed organic anion/cation transporter. OCTN2 plays a key role in absorption, tissue distribution and renal reabsorption of L-carnitine which is an essential cofactor in fat metabolism. Defects in the OCTN2 are responsible for primary carnitine deficiency. OCTN2 also transports some important respiratory medicines (such as ipratropium and tiotropium), and due to its expression in the lung, may influence the disposition and absorption of these medicines in the lung.

Western Blot: Mouse small intestine Tissue, 1:200-1:2000; IHC: Human Kidney Tissue, 1:20-1:200

Type: Primary
Antigen: OCTN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR