Anti-PEX19 Rabbit Polyclonal Antibody

Supplier: Proteintech
14713-1-AP-150UL
10091-458EA 462.47 USD
10091-458
Anti-PEX19 Rabbit Polyclonal Antibody
Antibodies
Peroxins (PEXs) are proteins that are essential for the assembly of functional peroxisomes. PEX19 gene is necessary for early peroxisomal biogenesis. It acts both as a cytosolic chaperone and as an import receptor for peroxisomal membrane proteins (PMPs) . PEX19 may bind newly synthesized PMPs and facilitate their insertion into the peroxisome membrane are a group of genetically heterogeneous autosomal recessive, lethal diseases characterized by multiple defects in peroxisome function. Defects in this gene are a cause of Zellweger syndrome (ZWS), as well as peroxisome biogenesis disorder complementation group 14 (PBD-CG14) .

Western Blot: Human Heart Tissue, 1:200-1:2000; IHC: Human gliomas Tissue, 1:20-1:200; IP:K-562 Cells, 1:200-1:1000

Type: Primary
Antigen: PEX19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR