Anti-NNT Rabbit Polyclonal Antibody

Supplier: Proteintech
13442-2-AP-150UL
10091-176EA 462.47 USD
10091-176
Anti-NNT Rabbit Polyclonal Antibody
Antibodies
NNT(nicotinamide nucleotide transhydrogenase) is a transmembrane protein and functions as a proton pumping transhydrogenase. The protein is present in both prokaryotes and eukaryotes and is located in the inner membrane of mitochondria. In prokaryotic cells, the enzyme is composed of α and βsubunits of 54 and 48 kDa, respectively. In eukaryotic cells, the enzyme is usually composed of a single peptide of 110 kDa. Although NNT catalyzes the interconversion of NADH and NADPH, the forward reaction using the reducing power of NADH to regenerate NADPH would be favored under conditions of oxidative stress. It can exsit as a homodimer.

Western Blot: Human adrenal gland Tissue, 1:200-1:2000; IHC: Human Liver cancer Tissue, 1:20-1:200

Type: Primary
Antigen: NNT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR