Anti-IFIT1 Rabbit Polyclonal Antibody

Supplier: Proteintech
23247-1-AP-150UL
10088-596EA 462.47 USD
10088-596
Anti-IFIT1 Rabbit Polyclonal Antibody
Antibodies
IFIT1 is, also known as glucocorticoid-attenuated response gene 16 protein (GARG-16), is a 463 amino acid protein belonging to the IFIT family. IFIT genes comprise a large family with three (Ifit1, Ifit2, and Ifit3) and four (IFIT1, IFIT2, IFIT3, and IFIT5) members in mice and humans, respectively. IFIT1 specifically bound virus PPP-RNA forms a complex with IFIT2 and IFIT3 to sequester the viral PPP-RNA and prevent virus replication.

IHC: Human breast cancer, 1:20-1:200; Western Blot:U-937 Cells, 1:200-1:2000

Type: Primary
Antigen: IFIT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR