Anti-GAD1 Rabbit Polyclonal Antibody

Supplier: Proteintech
10408-1-AP-150UL
10087-300EA 462.47 USD
10087-300
Anti-GAD1 Rabbit Polyclonal Antibody
Antibodies
The GAD1(glutamate decarboxylase 1) gene encodes 67 kDa glutamic acid decarboxylase isoform (GAD67), the rate-limiting enzyme responsible for γ-aminobutyric acid (GABA) biosynthesis from glutamatic acid and the major GAD isoform in the human brain for early brain development. It is also potentially involved in variety of skin activities. It belongs to the group II decarboxylase family.

Type: Primary
Antigen: GAD1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR