Anti-FYTTD1 Rabbit Polyclonal Antibody

Supplier: Proteintech
24560-1-AP-150UL
10087-206EA 462.47 USD
10087-206
Anti-FYTTD1 Rabbit Polyclonal Antibody
Antibodies
FYTTD1 also termed as UAP56 interacting factor or UIF is a 318 amino acid protein, which belongs to the UIF family. FYTTD1 localizes to nucleus and is required for mRNA export from nucleus to cytoplasm. Functioning as an adaptor, FYTTD1 utilizes the BAT1/DDX39-TAP pathway, which is essential for efficient mRNA export and nuclear pore delivery. FYTTD1 interacts with SSRP1, a protein that is necessary for its recruitment of mRNAs, in addition to a mutually exclusive interaction with BAT1/DDX39 and TAP. The molecular weight of post-translation modified FYTTD1 is observed 50 kDa.

Western Blot: HeLa Cells, 1:200-1:1000

Type: Primary
Antigen: FYTTD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR