Anti-FXC1 Rabbit Polyclonal Antibody

Supplier: Proteintech
10907-1-AP-150UL
10087-202EA 462.47 USD
10087-202
Anti-FXC1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FXC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR