Anti-FUT9 Mouse Monoclonal Antibody [clone: 5D4G6]

Supplier: Proteintech
60230-1-IG-150UL
10087-200EA 462.47 USD
10087-200
Anti-FUT9 Mouse Monoclonal Antibody [clone: 5D4G6]
Antibodies
Western Blot: Human Brain, 1:200-1:2000; IHC: Human Stomach Tissue, 1:20-1:200

Type: Primary
Antigen: FUT9
Clonality: Monoclonal
Clone: 5D4G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR