Anti-FUT8 Mouse Monoclonal Antibody [clone: 2F7H2]

Supplier: Proteintech
66118-1-IG-150UL
10087-196EA 462.47 USD
10087-196
Anti-FUT8 Mouse Monoclonal Antibody [clone: 2F7H2]
Antibodies
Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human lung cancer Tissue, 1:20-1:200

Type: Primary
Antigen: FUT8
Clonality: Monoclonal
Clone: 2F7H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR