Anti-FSTL1 Rabbit Polyclonal Antibody

Supplier: Proteintech
20182-1-AP-150UL
10087-166EA 462.47 USD
10087-166
Anti-FSTL1 Rabbit Polyclonal Antibody
Antibodies
Western Blot:A2780 Cells, 1:200-1:2000; IHC: Human pancreas Tissue, 1:20-1:200

Type: Primary
Antigen: FSTL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR