Anti-FPGT Rabbit Polyclonal Antibody

Supplier: Proteintech
24937-1-AP-150UL
10087-138EA 462.47 USD
10087-138
Anti-FPGT Rabbit Polyclonal Antibody
Antibodies
FPGT(Fucose-1-phosphate guanylyltransferase) is also named as GFPP. This protein catalyzes the formation of GDP-L-fucose from GTP and L-fucose-1-phosphate. It also functions as a salvage pathway to reutilize L-fucose arising from the turnover of glycoproteins and glycolipids. FPGT has 6 isoforms produced by alternative splicing.

Western Blot: Mouse Colon Tissue, 1:200-1:2000; IHC: Human Colon Tissue, 1:20-1:200

Type: Primary
Antigen: FPGT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR