Anti-FOXE3 Rabbit Polyclonal Antibody

Supplier: Proteintech
55301-1-AP-150UL
10087-114EA 462.47 USD
10087-114
Anti-FOXE3 Rabbit Polyclonal Antibody
Antibodies
FOXE3, also named as FKHL12 and FREAC8, is a forkhead transcription factor expressed in the lens which is located at chromosome 1p32, have been associated with both recessive and dominant ocular disease. Defects in FOXE3 are a cause of anterior segment mesenchymal dysgenesis (ASMD). Defects in FOXE3 are a cause of congenital primary aphakia (CPA). This antibody is specific to FOXE3.

Western Blot: HepG2 Cells, 1:200-1:2000

Type: Primary
Antigen: FOXE3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR