Anti-FNDC3B Rabbit Polyclonal Antibody

Supplier: Proteintech
22605-1-AP-150UL
10087-092EA 462.47 USD
10087-092
Anti-FNDC3B Rabbit Polyclonal Antibody
Antibodies
Western Blot:Jurkat Cells, 1:500-1:5000

Type: Primary
Antigen: FNDC3B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR