Anti-FMO2 Rabbit Polyclonal Antibody

Supplier: Proteintech
15725-1-AP-150UL
10087-074EA 462.47 USD
10087-074
Anti-FMO2 Rabbit Polyclonal Antibody
Antibodies
FMO2, belongs to FMO (flavin containing monooxygenase) family, which catalyzes the oxidation of soft nucleophilic heteroatom centers in drugs, pesticides, and xenobiotics. FMO family has 1-5 members, with homologous sequences. This antibody is specially against FMO2. It does not bind other FMOs.

Western Blot: Mouse lung Tissue, 1:200-1:2000; IHC: Human lung cancer, 1:20-1:200

Type: Primary
Antigen: FMO2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR