Anti-FN1 Mouse Monoclonal Antibody [clone: 1G10F9]

Supplier: Proteintech
66042-1-IG-150UL
10087-016EA 462.47 USD
10087-016
Anti-FN1 Mouse Monoclonal Antibody [clone: 1G10F9]
Antibodies
Fibronectin 1 (FN1) is a high molecular weight glycoprotein which exists in both a soluble form in plasma (plasma FN1) and other body fluids and an insoluble form in the extracellular matrix (cellular FN1). Plasma FN1 (dimeric form) is secreted by hepatocytes. Cellular FN (dimeric or cross-linked multimeric forms), made by fibroblasts, epithelial and other cell types, is deposited as fibrils in the extracellular matrix. FN1 binds to cell surfaces through integrins and to various compounds including collagen, fibrin and heparin. It is involved in cell adhesion and migration processes including embryogenesis, wound healing, hemostasis, host defense, and metastasis.

Western Blot: Human Brain, 1:200-1:2000

Type: Primary
Antigen: Fibronectin
Clonality: Monoclonal
Clone: 1G10F9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR