Anti-FHL1 Rabbit Polyclonal Antibody

Supplier: Proteintech
51015-1-AP-150UL
10087-004EA 462.47 USD
10087-004
Anti-FHL1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Heart Tissue, 1:500-1:5000; IHC: Human ovary tumor, 1:20-1:200

Type: Primary
Antigen: Fhl1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR