Anti-Fibrinogen gamma chain Rabbit Polyclonal Antibody

Supplier: Proteintech
15841-1-AP-150UL
10086-994EA 462.47 USD
10086-994
Anti-Fibrinogen gamma chain Rabbit Polyclonal Antibody
Antibodies
Fibrinogen is a soluble plasma glycoprotein synthesized in the liver. It is composed of two sets of three structurally different subunits: alpha (FGA), beta (FGB), gamma (FGG). Fibrinogen is converted by thrombin into fibrin during blood coagulation. Fibrinogen and fibrin play overlapping roles in blood clotting, fibrinolysis, cellular and matrix interactions, the inflammatory response, wound healing, and neoplasia . FGG is the gamma chain of fibrinogen. Mutations in the gene of FGG lead to several disorders, including dysfibrinogenemia, hypofibrinogenemia and thrombophilia.

Western Blot: Human spleen Tissue, 1:500-1:5000; IHC: Human hepatocirrhosis Tissue, 1:20-1:200

Type: Primary
Antigen: Fibrinogen gamma chain
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR