Anti-Fibrinogen gamma chain Mouse Monoclonal Antibody [clone: 2B2B8]
Supplier: Proteintech
66158-1-IG-150UL
10086-992EA 462.47 USD
10086-992
Anti-Fibrinogen gamma chain Mouse Monoclonal Antibody [clone: 2B2B8]
Antibodies
Fibrinogen is a soluble plasma glycoprotein synthesized in the liver. It is composed of two sets of three structurally different subunits: alpha (FGA), beta (FGB), gamma (FGG). Fibrinogen is converted by thrombin into fibrin during blood coagulation. Fibrinogen and fibrin play overlapping roles in blood clotting, fibrinolysis, cellular and matrix interactions, the inflammatory response, wound healing, and neoplasia . FGG is the gamma chain of fibrinogen. Mutations in the gene of FGG lead to several disorders, including dysfibrinogenemia, hypofibrinogenemia and thrombophilia.
Western Blot:More Product Information