Anti-FBXO41 Rabbit Polyclonal Antibody

Supplier: Proteintech
24519-1-AP-150UL
10086-914EA 462.47 USD
10086-914
Anti-FBXO41 Rabbit Polyclonal Antibody
Antibodies
FBXO41 also named as F box only protein 41 is a 875 amino-acid protein, which directly interact with SKP1 and CUL1. FBXO41 is identified as a Substrate-recognition component of the SCF (SKP1-CUL1-F-box protein)-type E3 ubiquitin ligase complex, which as a regulator mediates some protein ubiquitination degradation. FBXO41 consists of F-box domain and has zinc ion binding activity. FBXO41 may have an association with Parkinson’s disease.

Western Blot: Mouse Brain Tissue, 1:500-1:5000; IP: Mouse Brain Tissue, 1:500-1:5000; IHC: Human Heart Tissue, 1:20-1:200

Type: Primary
Antigen: FBXO41
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR