Anti-FBLIM1 Rabbit Polyclonal Antibody

Supplier: Proteintech
13349-1-AP-150UL
10086-890EA 462.47 USD
10086-890
Anti-FBLIM1 Rabbit Polyclonal Antibody
Antibodies
FBLIM1, also known as CAL, Mig-2-interacting protein or Migfilin, is a cytoplasmic protein that belongs to the LIM superfamily. FBLIM1 is a protein found in cell-cell and cell-ECM connections where it co-localizes with FLNA/C and FLNB. FBLIM1 was found to bind directly to FLNA/C and to be an important regulator of cell shape and motility. FBLIM1 exerts its influence on cellular functions by interacting with various binding partners; FLN via its N- terminal domain, VASP and Src via its proline-rich region, and kindlin-2 and the cardiac transcription factor, CSX/NKX2-5 via its C-terminal LIM domains. Three isoforms exist for FBLIM1 due to alternative splicing events, namely FBLP-1A, FBLP-1 and FBLP-1B. FBLIM1 serves as an anchoring site for cell-ECM adhesion proteins and filamin-containing actin filaments. It is associated with actin stress fiber at cell-ECM focal adhesion sites. FBLP-1A and FBLP-1B are recruited and localized at actin stress fibers and clustered at cell-EMC adhesion sites through interaction with PLEKHC1. FBLP-1 is localized at actin stress fibers. FBLIM1 is implicated in cell shape modulation (spreading) and motility. FBLIM1 participate in the regulation of filamin-mediated cross-linking and stabilization of actin filaments. It may also regulate the assembly of filamin-containing signaling complexes that control actin assembly. In addition, FBLIM1 is capable of translocating to the nucleus and regulating gene expression. This antibody is a rabbit polyclonal antibody raised against full length human FBLIM1 antigen.

Western Blot: HepG2 Cells, 1:500-1:5000; IHC: Human lung cancer, 1:20-1:200

Type: Primary
Antigen: FBLIM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR