Anti-FANCI Rabbit Polyclonal Antibody

Supplier: Proteintech
20789-1-AP-150UL
10086-860EA 462.47 USD
10086-860
Anti-FANCI Rabbit Polyclonal Antibody
Antibodies
Western Blot: HepG2 Cells, 1:1000-1:10000; IF: Hela Cells, 1:10-1:100

Type: Primary
Antigen: FANCI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR