Anti-FANCG Rabbit Polyclonal Antibody

Supplier: Proteintech
10215-1-AP-150UL
10086-858EA 462.47 USD
10086-858
Anti-FANCG Rabbit Polyclonal Antibody
Antibodies
Western Blot:Jurkat Cells, 1:200-1:1000; IF: HeLa Cells, 1:10-1:100

Type: Primary
Antigen: FANCG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR