Anti-FAM98B Rabbit Polyclonal Antibody

Supplier: Proteintech
22251-1-AP-150UL
10086-846EA 462.47 USD
10086-846
Anti-FAM98B Rabbit Polyclonal Antibody
Antibodies
Western Blot: Raji Cells, 1:500-1:5000; IHC: Human breast cancer Tissue, 1:20-1:200

Type: Primary
Antigen: FAM98B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR