Anti-FAM3C Rabbit Polyclonal Antibody

Supplier: Proteintech
14247-1-AP-150UL
10086-816EA 462.47 USD
10086-816
Anti-FAM3C Rabbit Polyclonal Antibody
Antibodies
FAM3C, also known as ILEI, is a secreted protein belonging to the cytokine-like protein family. FAM3C is a signaling cytokine with a GG domain, hydrophobic leader sequence and an N-terminal signal peptide. A change in expression of this protein has been noted in pancreatic cancer-derived cells. FAM3C has been involved in the epithelial to mesenchymal transition, and retinal laminar formation processes in vertebrates. Also, FAM3C may play an important role in inner ear development and thus be involved in cell differentiation and proliferation during inner ear embryogenesis.

Western Blot: Human Brain Tissue, 1:200-1:2000; IHC: Human pancreas, 1:20-1:200

Type: Primary
Antigen: FAM3C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR