Anti-EYA3 Rabbit Polyclonal Antibody

Supplier: Proteintech
21196-1-AP-150UL
10086-740EA 462.47 USD
10086-740
Anti-EYA3 Rabbit Polyclonal Antibody
Antibodies
EYA3(Eyes absent homolog 3) belongs to the HAD-like hydrolase superfamily.Its function as histone phosphatase probably explains its role in transcription regulation during organogenesis.EYA3, like EYA1, is expressed in the ninth week of human development and may also underlie developmental defects.It has 2 isoforms produced by alternative splicing.This antibody is specific to EYA3.

Western Blot:L02 Cells, 1:500-1:5000; IF: Hela Cells, 1:10-1:100; IHC: Human breast cancer Tissue, 1:20-1:200

Type: Primary
Antigen: EYA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR