Anti-ETV5 Rabbit Polyclonal Antibody

Supplier: Proteintech
13011-1-AP-150UL
10086-686EA 462.47 USD
10086-686
Anti-ETV5 Rabbit Polyclonal Antibody
Antibodies
ETS variant gene 5(ETV5) belongs to the ETS oncogene family that shares a conserved peptide, ETS domain, which mediates sequence-specific DNA binding. ETV5 is a transcription factor and is required for spermatogonial stem cell self-renewal. ETV5 can be negatively regulated by COP1, a tumor suppressor. Also it has a role in branching morphogenesis in the developing kidney

Western Blot:PC-3 Cells, 1:500-1:5000

Type: Primary
Antigen: ETV5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR