Anti-ENOX2 Rabbit Polyclonal Antibody

Supplier: Proteintech
10423-1-AP-150UL
10086-534EA 462.47 USD
10086-534
Anti-ENOX2 Rabbit Polyclonal Antibody
Antibodies
Tumor-associated NADH oxidase (ENOX2) as a member of a family of growth-related NADH (or hydroquinone) oxidases. It functions as a terminal oxidase of plasma electron transport from cytosolic NAD(P)H via hydroquinones to acceptors at the cell surface. Hydroquinone oxidase activity alternates with a protein disulfide-thiol interchange/oxidoreductase activity which may control physical membrane displacements associated with vesicle budding or cell enlargement. The activities oscillate with a period length of 22 minutes and play a role in control of the ultradian cellular biological clock

IHC: Human ovary tumor Tissue, 1:20-1:200; Western Blot: MCF-7 Cells, 1:500-1:5000

Type: Primary
Antigen: ENOX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR