Anti-EIF1 Rabbit Polyclonal Antibody

Supplier: Proteintech
15276-1-AP-150UL
10086-334EA 462.47 USD
10086-334
Anti-EIF1 Rabbit Polyclonal Antibody
Antibodies
It is a necessary for scanning and involved in initiation site selection. Promotes the assembly of 48S ribosomal complexes at the authentic initiation codon of a conventional capped mRNA. Molecular weight is 13kd.

IHC: Human Prostate Cancer Tissue, 1:20-1:200

Type: Primary
Antigen: EIF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR