Anti-EEF1E1 Rabbit Polyclonal Antibody

Supplier: Proteintech
10805-1-AP-150UL
10086-276EA 462.47 USD
10086-276
Anti-EEF1E1 Rabbit Polyclonal Antibody
Antibodies
During the translation processes, aminoacyl-tRNA synthetases (ARSs) ligate specific amino acids to tRNAs. Together with non-enzymatic proteins called ARS-interacting multifunctional proteins (AIMPs), the ARSs form a macromolecular complex . This complex has an essential role in protein synthesis, but also in other biological processes including angiogenesis, apoptosis and inflammation. AIMP3 is not only associated with the multi-tRNA synthetase complex via its specific interaction with methionyl-tRNA synthetase, but also works as a tumor suppressor by interacting with ATM, the upstream kinase of p53 .

Western Blot: Human Testis Tissue, 1:200-1:2000; IHC: Human breast cancer, 1:20-1:200

Type: Primary
Antigen: EEF1E1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR