Anti-ECEL1 Rabbit Polyclonal Antibody

Supplier: Proteintech
14222-1-AP-150UL
10086-240EA 462.47 USD
10086-240
Anti-ECEL1 Rabbit Polyclonal Antibody
Antibodies
The M13 family has seven members, including ECEL1 (endothelin-converting enzyme-like 1), one of the newest members. ECEL1 is expressed predominantly in the central nervous system. It has been proposed that the enzyme has a role in the nervous regulation of the respiratory system.It may contribute to the degradation of peptide hormones and be involved in the inactivation of neuronal peptides.It has 2 isoforms produced by alternative splicing and 3 glycosylation sites.It can be detected a band of 95kDa in CHO cells by western blot because of the heavily glycosylated.

Type: Primary
Antigen: ECEL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR