Anti-DYNC1I2 Rabbit Polyclonal Antibody

Supplier: Proteintech
12219-1-AP-150UL
10086-206EA 462.47 USD
10086-206
Anti-DYNC1I2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Brain Tissue, 1:500-1:5000; IF:A375 Cells, 1:10-1:100

Type: Primary
Antigen: DYNC1I2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR