Anti-DSE Mouse Monoclonal Antibody [clone: 2C9F7]

Supplier: Proteintech
60041-1-IG-150UL
10086-138EA 462.47 USD
10086-138
Anti-DSE Mouse Monoclonal Antibody [clone: 2C9F7]
Antibodies
DSE, also named as SART2 and DSEPI, is an enzyme that converts D-glucuronic acid to L-iduronic acid residues in dermatan sulphate biosynthesis. It is also identified to be a tumour-associated antigen. DSE is recognized by cytotoxic T cells (CTLs) and its enhanced expression in many cancers has been reported. DSE is a potential candidate for a tumour antigen with immunogenicity.

Western Blot:COLO 320 Cells, 1:200-1:1000

Type: Primary
Antigen: DSE
Clonality: Monoclonal
Clone: 2C9F7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR