Anti-DR3 Rabbit Polyclonal Antibody

Supplier: Proteintech
55397-1-AP-150UL
10086-110EA 462.47 USD
10086-110
Anti-DR3 Rabbit Polyclonal Antibody
Antibodies
TNFRSF25, also named as APO3, DDR3, DR3, TNFRSF12, WSL, WSL1, LARD, TRAMP and AIR, is a receptor for TNFSF12/APO3L/TWEAK. TNFRSF25 interacts directly with the adapter TRADD. It mediates activation of NF-kappa-B and induces apoptosis. TNFRSF25 may play a role in regulating lymphocyte homeostasis. It is constitutively and highly expressed by CD4(+)FoxP3(+) Tregs. This antibody can recognize all the isoform of TNFRSF25.

Western Blot:PC-3 Cells, 1:200-1:2000

Type: Primary
Antigen: DR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR