Anti-DOCK9 Rabbit Polyclonal Antibody

Supplier: Proteintech
18987-1-AP-150UL
10086-068EA 462.47 USD
10086-068
Anti-DOCK9 Rabbit Polyclonal Antibody
Antibodies
DOCK9 is a guanine nucleotide exchange factor (GEF) that activates CDC42 by exchanging bound GDP for free GTP. Its overexpression induces filopodia formation.

Western Blot:A549 Cells, 1:200-1:2000; IHC: Human Heart, 1:20-1:200; IF: Hela, 1:10-1:100

Type: Primary
Antigen: DOCK9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR