Anti-DHPS Rabbit Polyclonal Antibody

Supplier: Proteintech
11184-1-AP-150UL
10085-816EA 462.47 USD
10085-816
Anti-DHPS Rabbit Polyclonal Antibody
Antibodies
DHPS(Deoxyhypusine synthase) catalyzes the conversion of lysine to deoxyhypusine residue on the eukaryotic initiation factor 5A (eIF-5A) precursor using spermidine as the substrate. It belongs to the deoxyhypusine synthase family. This protein has 3 isoforms produced by alternative splicing.

Western Blot: HeLa Cells, 1:200-1:1000

Type: Primary
Antigen: DHPS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR