Anti-DHODH Rabbit Polyclonal Antibody

Supplier: Proteintech
14877-1-AP-150UL
10085-814EA 462.47 USD
10085-814
Anti-DHODH Rabbit Polyclonal Antibody
Antibodies
DHODH(Dihydroorotate dehydrogenase) catalyzes the fourth enzymatic step in de novo pyrimidine biosynthesis.DHO dehydrogenase is a monofunctional protein which, in most eukaryotic organisms, is located on the outer surface of the inner mitochondrial membrane.Defects in DHODH are the cause of postaxial acrofacial dysostosis (POADS).

Western Blot: MCF7 Cells, 1:500-1:5000; IHC: Human breast cancer Tissue, 1:20-1:200; IF: MCF-7 Cells, 1:20-1:200

Type: Primary
Antigen: DHODH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR