Anti-DMRTC1B Rabbit Polyclonal Antibody

Supplier: Proteintech
21473-1-AP-150UL
10085-574EA 462.47 USD
10085-574
Anti-DMRTC1B Rabbit Polyclonal Antibody
Antibodies
Western Blot: MCF7 Cells, 1:200-1:2000

Type: Primary
Antigen: DMRTC1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR