Anti-DMKN Rabbit Polyclonal Antibody

Supplier: Proteintech
16252-1-AP-150UL
10085-572EA 462.47 USD
10085-572
Anti-DMKN Rabbit Polyclonal Antibody
Antibodies
DMKN, also named as epidermis-specific secreted protein SK30/SK89, acts as a soluble regulator of keratinocyte differentiation. DMKN has 16 isoforms with MW 9-20 kDa, 35-38 kDa and 42-48 kDa.

Western Blot: Mouse ovary Tissue, 1:500-1:5000; IHC: Human skin Tissue, 1:20-1:200

Type: Primary
Antigen: DMKN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR