Anti-DDX50 Rabbit Polyclonal Antibody

Supplier: Proteintech
10358-1-AP-150UL
10085-570EA 462.47 USD
10085-570
Anti-DDX50 Rabbit Polyclonal Antibody
Antibodies
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this DEAD box protein family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. DEAD box polypeptide 50 (DDX50, synonyms: GU2, GUB, MGC3199, RH-II/GuB) enzyme may be involved in ribosomal RNA synthesis or processing.

Western Blot: HeLa Cells, 1:200-1:2000; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: DDX50
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR