Anti-CYP4A11 Rabbit Polyclonal Antibody

Supplier: Proteintech
11688-1-AP-150UL
10085-538EA 462.47 USD
10085-538
Anti-CYP4A11 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Kidney Tissue, 1:200-1:2000; IHC: Human Kidney, 1:20-1:200

Type: Primary
Antigen: CYP4A11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR