Anti-CYP1A1 Rabbit Polyclonal Antibody

Supplier: Proteintech
13241-1-AP-150UL
10085-502EA 462.47 USD
10085-502
Anti-CYP1A1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse cerebellum Tissue, 1:500-1:5000; IHC: Human breast cancer Tissue, 1:20-1:200; FC:Jurkat Cells, N/A

Type: Primary
Antigen: CYP1A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR