Anti-CXCR7 Mouse Monoclonal Antibody [clone: 4C3D7]

Supplier: Proteintech
60216-1-IG-150UL
10085-436EA 462.47 USD
10085-436
Anti-CXCR7 Mouse Monoclonal Antibody [clone: 4C3D7]
Antibodies
CXCR7 (C-X-C chemokine receptor type 7), also known as RDC1, is a member of the G-protein coupled receptor family. CXCR7 can bind the chemokines CXCL11 and CXCL12 with high affinity, and it also acts as coreceptor with CXCR4 for a restricted number of HIV isolates. Expression of CXCR7 has been associated with cardiac development as well as with tumor growth and progression.

Western Blot:Jurkat Cells, 1:500-1:5000

Type: Primary
Antigen: CXCR7
Clonality: Monoclonal
Clone: 4C3D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR