Anti-CTNNBL1 Rabbit Polyclonal Antibody

Supplier: Proteintech
13665-1-AP-150UL
10085-318EA 462.47 USD
10085-318
Anti-CTNNBL1 Rabbit Polyclonal Antibody
Antibodies
Western Blot:Jurkat Cells, 1:500-1:5000; IHC: Human Testis Tissue, 1:20-1:200

Type: Primary
Antigen: CTNNBL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR