Anti-CSTF1 Rabbit Polyclonal Antibody

Supplier: Proteintech
15537-1-AP-150UL
10085-278EA 462.47 USD
10085-278
Anti-CSTF1 Rabbit Polyclonal Antibody
Antibodies
Cleavage stimulation factor (CstF) , a heterotrimeric protein with subunits of 77, 64, and 50 kD (CstF-77, -64, and -50), is a polyadenylation factor that helps to specify the site of processing. CstF recognizes the G+U-rich element, a sequence located downstream of the cleavage site. CstF-50 unit interacts with the COOH-terminal domain of the RNAP II largest subunit (CTD).

Western Blot: HeLa Cells, 1:500-1:5000; IP:K-562 Cells, 1:200-1:2000

Type: Primary
Antigen: CSTF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR