Anti-CSRP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
13432-1-AP-150UL
10085-264EA 462.47 USD
10085-264
Anti-CSRP1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Stomach Tissue, 1:200-1:2000; IHC: Human cervical cancer, 1:20-1:200

Type: Primary
Antigen: CSRP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR