Anti-CRX Rabbit Polyclonal Antibody

Supplier: Proteintech
12047-1-AP-150UL
10085-216EA 462.47 USD
10085-216
Anti-CRX Rabbit Polyclonal Antibody
Antibodies
Cone-rod homeobox (Crx) is a homeodomain transcription factor and member of the Otx family, which has been thought to play a critical role in determining and maintaining the phenotype of both pinealocytes and retinal photoreceptors . In the mammalian retina, Crx plays an essential role in the normal development and maintenance of cones and rods and regulates expression of the network of genes that characterize the retina . Elimination of Crx by disruption of the homeobox domain results in loss of the image-forming visual system but not the non-image-forming visual system controlling circadian rhythms .

Western Blot:Y79 Cells, 1:200-1:2000

Type: Primary
Antigen: CRX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR