Anti-COX5A Rabbit Polyclonal Antibody

Supplier: Proteintech
11448-1-AP-150UL
10085-060EA 462.47 USD
10085-060
Anti-COX5A Rabbit Polyclonal Antibody
Antibodies
COX5A(Cytochrome c oxidase subunit 5A,mitochondrial)is also named as COXVA and belongs to the cytochrome c oxidase subunit 5A family.This is the heme A-containing chain of cytochrome c oxidase, the terminal oxidase in mitochondrial electron transport.The mature human COX Va polypeptide shares 35% and 95% protein sequence identity with the corresponding S. cerevisiae COX subunit VI and bovine COX Va, respectively.

IHC: Human gliomas Tissue, 1:20-1:200; Western Blot: HeLa Cells, 1:200-1:2000

Type: Primary
Antigen: COX5A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR