Anti-CORO1C Rabbit Polyclonal Antibody

Supplier: Proteintech
14749-1-AP-150UL
10085-032EA 462.47 USD
10085-032
Anti-CORO1C Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Skeletal Muscle Tissue, 1:500-1:5000; IHC: Human cervical cancer, 1:20-1:200; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: CORO1C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR