Anti-CLSTN1 Rabbit Polyclonal Antibody

Supplier: Proteintech
12788-1-AP-150UL
10084-880EA 462.47 USD
10084-880
Anti-CLSTN1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Brain Tissue, 1:500-1:5000

Type: Primary
Antigen: CLSTN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR